Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon Dioxide, 325 Mesh, FCC, 90.4%, Spectrum™ Chemical
SDP

Catalog No. 18606217 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
20 kg
Packaging:
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-217 2.5 kg Poly Pail
18-606-218 12 kg Fiber Drum
18-606-219 20 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-606-217 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S13882.5KG
Add to Cart
Add to Cart

S1388, 7631-86-9, SiO2·xH2O

Spectrum™ Chemical Silicon Dioxide, 325 Mesh, FCC is used as a food additive as a powder flow helper. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

CAS Max % 1
Material Silicon Dioxide
Quantity 2.5 kg
Loss on Drying 0.07
Loss on Ignition 0.085
Mesh Size 325
Packaging Poly Pail
Percent Purity 94%
Grade FCC
Chemical Name or Material Silicon Dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.