Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon(IV) 2-ethylhexanoate, nominally 75% in ethanol

Catalog No. aa4463530
Click to view available options
Quantity:
250 g

CAS: 67939-81-5 | C32H60O8Si | 600.909 g/mol

In the manufacture of its esters for artificial flavors, and in the manufacture of hexyl derivatives, such as hexylphenols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67939-81-5
Molecular Formula C32H60O8Si
Molecular Weight (g/mol) 600.909
MDL Number MFCD00269842
InChI Key UNKOLEUDCQIVBV-UHFFFAOYSA-N
Synonym silicon 2-ethylhexanoate, tetrakis 2-ethylhexanoic acid, tetraanhydride with silicic acid, 2-ethylhexanoic acid tetraanhydride with silicic acid h4sio4, hexanoic acid, 2-ethyl-, 1,1',1,1'-tetraanhydride with silicic acid h4sio4, hexanoic acid, 2-ethyl-, tetraanhydride with silicic acid h4sio4, silicon 2-ethylhexanoate, min. 90, tris 2-ethylhexanoyloxy silyl 2-ethylhexanoate, silicon iv 2-ethylhexanoate in ethanol, silicic acid tetrakis 2-ethylhexanoic tetraanhydride, silicon iv 2-ethylhexanoate, nominally in ethanol
PubChem CID 106984
IUPAC Name tris(2-ethylhexanoyloxy)silyl 2-ethylhexanoate
SMILES CCCCC(CC)C(=O)O[Si](OC(=O)C(CC)CCCC)(OC(=O)C(CC)CCCC)OC(=O)C(CC)CCCC

Specifications

Color Yellow
Quantity 250 g
UN Number UN1170
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 600.91
Physical Form Liquid
Chemical Name or Material Silicon (IV) 2-ethylhexanoate
Hazard Category H225-H361d
Hazard Statement GHS H Statement
H225-H361
Highly flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P281-P303+P361+P353-P308+P313-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS
TSCA Yes
Recommended Storage Ambient temperatures

For Manufacturing and Laboratory Use. Not for beverage use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.