Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon(IV) bromide

Catalog No. p-7049333
Click to view available options
Quantity:
10 g
50 g

CAS: 7789-66-4 | Br4Si | 347.701 g/mol

Silicon(IV) bromide is used to grow thin films of silicon by the chemical vapor deposition method. It is also used for ceramics, sealants and the production of many cutting tools.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-66-4
Molecular Formula Br4Si
Molecular Weight (g/mol) 347.701
MDL Number MFCD00049530
InChI Key AIFMYMZGQVTROK-UHFFFAOYSA-N
Synonym silicon iv bromide, silicon tetrabromide, silane, tetrabromo, sibr4, unii-8gac9ywl42, 8gac9ywl42, silicon bromide, acmc-20ajqj
PubChem CID 82247
IUPAC Name tetrabromosilane
SMILES [Si](Br)(Br)(Br)Br

Specifications

Boiling Point 154°C
Melting Point 5°C
Color Colorless to Yellow
Physical Form Liquid
Quantity 10 g
UN Number UN3264
Merck Index 14,8499
Formula Weight 347.72
Odor Stench
Sensitivity Moisture sensitive
Density 2.800 g/mL
Chemical Name or Material Silicon(IV) bromide
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 232-182-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.