Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Silicon(IV) chloride, 99.8+%
Click to view available options
Quantity:
50 mL
500 mL
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 58.0°C |
Melting Point | -70.0°C |
Color | Colorless |
Physical Form | Liquid |
Quantity | 2.5 L |
Assay Percent Range | 99.8+% |
Linear Formula | SiCl4 |
Fieser | 10,347; 11,466; 16,317 |
Merck Index | 15, 8639 |
Solubility Information | Solubility in water: decomposes. Other solubilities: miscible with benzene,ether,chloroform,,petroleum ether,carbon tetrachloride,,tin tetrachloride,sulfur,mono- and dichlorides,decomposes by alcohol with evolution of hydrogen,chloride |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement May cause respiratory irritation. Toxic if swallowed. Toxic if inhaled. Causes severe skin burns and eye damage. Reacts violently with water. |
Precautionary Statement | GHS P Statement Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Avoid breathing dust/fume/gas/mist/vapors/spray. |
EINECSNumber | 233-054-0 |
RTECSNumber | VW0525000 |
TSCA | TSCA |
Recommended Storage | Product may develop some turbidity or precipitate |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction