Alfa Aesar™ Silicon(IV) iodide, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4414122

Quantity 100g

 View more versions of this product

Catalog No. AA4414122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13465-84-4
    Molecular Formula I4Si
    Linear Formula SiI4
    Formula Weight 535.7
    MDL Number MFCD00054041
    UN Number UN2923
    Chemical Name or Material Silicon (IV) iodide

    Specifications

    Formula Weight 535.7
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Sensitivity Light, Air & Moisture Sensitive
    Molecular Formula I4Si
    MDL Number MFCD00054041
    Boiling Point 290°C
    Quantity 100g
    Linear Formula SiI4
    CAS 13465-84-4
    Color White
    Melting Point 120°C
    Density 4.198g/mL
    Physical Form Lumps
    UN Number UN2923
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H314-H318
    Fatal if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Storage Note 1
    • Packaged under Argon
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
    Chemical Name or Material Silicon (IV) iodide
    Assay 99.999%, (metals basis)

    GHS H Statement
    H300-H314-H318
    Fatal if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit