Silicon(IV) iodide, 99% (metals basis), Alfa Aesar™

Light, Air and Moisture Sensitive

Manufacturer: Alfa Aesar 8730306

Quantity 5g

 View more versions of this product

Catalog No. AA8730306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13465-84-4
    Physical Form Crystals, Packaged Under Argon
    Assay Percent Range 99% (metals basis)
    Molecular Formula I4Si
    Formula Weight 535.7
    MDL Number MFCD00054041
    Synonym Silicon tetraiodide
    Chemical Name or Material Silicon(IV) iodide

    Specifications

    Physical Form Crystals, Packaged Under Argon
    Formula Weight 535.7
    Quantity 5g
    Color White
    DOT Information DOT Class: 8, Packaging Group: II
    Density 4.198g/cm3 at 20°C
    EINECS Number 236-706-2
    Assay Percent Range 99% (metals basis)
    Molecular Formula I4Si
    MDL Number MFCD00054041
    CAS 13465-84-4
    Synonym Silicon tetraiodide
    Health Hazard 1 H300-H314-H318: Fatal if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage.
    Chemical Name or Material Silicon(IV) iodide

    H300-H314-H318: Fatal if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit