Learn More
Silicon(IV) oxide, 99.0% (metals basis)
Description
Silicon(IV) oxide is used in the manufacture of glass for windows, drinking glasses, beverage bottles and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 2230°C |
Melting Point | 1710°C |
Physical Form | Fused Amorphous Powder |
Quantity | 1 kg |
Assay Percent Range | 99.8% (metals basis) |
Solubility Information | Insoluble in water. |
IUPAC Name | dioxosilane |
Formula Weight | 60.09 |
Density | 2.2 to 2.6 g/mL |
Chemical Name or Material | Silicon(IV) oxide |
Safety and Handling
EINECSNumber | 231-545-4 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.