Learn More
Silicon(IV) oxide, amorphous fumed, S.A. 130-170m2/g, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Boiling Point | 2230°C |
| Melting Point | 1710°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 250 g |
| Linear Formula | SiO2 |
| Merck Index | 14,8493 |
| Solubility Information | Insoluble in water. |
| IUPAC Name | silanedione |
| Formula Weight | 60.09 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | Warning |
| missing translation for 'healthHazard2' | GHS H Statement H373 May cause damage to organs through prolonged or repeated exposure. |
| missing translation for 'healthHazard3' | GHS P Statement P260-P314-P501a Do not breathe dust/fume/gas/mist/vapors/spray. Get medical advice/attention if you feel unwell. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 231-545-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.