Silicon powder, crystalline, -100 mesh, 99.9% (metals basis), Alfa Aesar

Manufacturer: Alfa Aesar 0031122

Quantity 100g

 View more versions of this product

Catalog No. AA0031122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-21-3
    Physical Form Crystalline Powder
    Assay Percent Range 99.9% (Metals basis)
    Molecular Formula Si
    Formula Weight 28.0855
    MDL Number MFCD00085311
    Chemical Name or Material Silicon

    Specifications

    Physical Form Crystalline Powder
    Formula Weight 28.0855
    Quantity 100g
    DOT Information DOT Class: 4.1, Packaging Group: III
    Density 2.33g/cm3
    EINECS Number 231-130-8
    Odor Odorless
    Assay Percent Range 99.9% (Metals basis)
    Appearance Powder
    MDL Number MFCD00085311
    Health Hazard 1 H228-H303: Flammable solid. May be harmful if swallowed.
    Molecular Formula Si
    CAS 7440-21-3
    Chemical Name or Material Silicon

    H228-H303: Flammable solid. May be harmful if swallowed.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit