Learn More
Cytiva Whatman™ O-Ring for use with MV050 Vacuum Filtration Manifolds
Catalog No. 01184190 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
MV050 Manifold Lids
Catalog No. For Use With (Equipment)
01-184-190 MV050 Manifold Lids
1 options
Catalog No. 01-184-190 Supplier Cytiva Supplier No. 10440006
Only null left
Add to Cart
Add to Cart
Replacement silicone O-ring for use with MV050 Vacuum Filtration Manifolds.
  • Product Type: O-Ring
  • Material: Silicone
  • For Use with (Equipment): MV050 Vacuum Filtration Manifold Lids
  • For Use with (Applications): Microbiology, Biochemistry, and Hydrobiology. Residue Analysis, Precipitate Analysis, and Contamination Testing (Food and Beverage, Pharmaceutical, and Cosmetic Industries)
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Disposable Reusable
For Use With (Equipment) MV050 Manifold Lids
Material Silicone
Sterility Non-sterile
For Use With (Application) Microbiology, Biochemistry, and Hydrobiology. Residue Analysis, Precipitate Analysis, and Contamination Testing (Food and Beverage, Pharmaceutical, and Cosmetic Industries)
Model MV050/0/09
Quantity 1 Ea.
Series MV050
Type O-Ring