Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Silicone oil
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
250 mL
1 L
Description
Silicone oil is a liquid based siloxane that is part of the methyl silicone fluid system. It has a viscosity of 500 cSt that is dependent on the chain length distribution. Its refractive index is ∼ 1.402 and dielectric strength is ∼ 14 kV/mm. It's surface tension tends to increase with an increase in the viscosity.Silicone oil can be used for a variety of application such as: heat transferring medium in chemical and petrochemical industries, a dielectric coolant, a lubricant and antiflatulent agent, protective coatings for construction materials, a cosmetic additive.
Specifications
Specifications
| CAS | 63148-62-9 |
| Density | 0.97 g/mL (at 25°C) |
| Boiling Point | >140°C (0.002 mmHg) |
| Linear Formula | [-Si(CH3)2 O-]n |
| Refractive Index | n20/D 1.403 (literature) |
| MDL Number | MFCD00132673 |
| Quantity | 1 L |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction