Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Silicone oil
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
250 mL
1 L
Description
Silicone oil is a liquid based siloxane that is part of the methyl silicone fluid system. It has a viscosity of 10000 cSt with a refractive index of ∼ 1.404 and a dielectric strength of ∼ 15 kV/mm. It's surface tension tends to increase with an increase in the viscosity.Silicone oil can be used for applications such as: protective coatings for building materials, a cosmetic additive, a dielectric coolant, a lubricant and antiflatulent agent.
Specifications
Specifications
| CAS | 63148-62-9 |
| Density | 0.971 g/mL (at 25°C) |
| Boiling Point | >140°C (0.002 mmHg) |
| Linear Formula | [-Si(CH3)2 O-]n |
| Refractive Index | n20/D 1.403 (literature) |
| Quantity | 250 mL |
| MDL Number | MFCD00132673 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction