Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicone oil, for oil baths, usable range from -40 to +200°C

Catalog No. AAA1272822
Click to view available options
Quantity:
100 g
500 g
2500 g

Catalog No. AAA1272822

Supplier: Thermo Scientific Chemicals A1272822

Only null left
Add to Cart

For oil baths | CAS: 63148-62-9 | (C2H6OSi)n | NaN g/mol

Silicone oil is used as a calibration media in temperature controlled baths in laboratories due to its good thermal characteristics and low flammability. It is also used as active component in silicone grease, lubricants and heat transfer fluids. It finds application in cosmetics, hair conditioners and as a sorbent for the analysis of headspace (dissolved gas analysis) of food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silicone oil is used as a calibration media in temperature controlled baths in laboratories due to its good thermal characteristics and low flammability. It is also used as active component in silicone grease, lubricants and heat transfer fluids. It finds application in cosmetics, hair conditioners and as a sorbent for the analysis of headspace (dissolved gas analysis) of food.

Solubility
Miscible with aliphatic, aromatic hydrocarbons, alcohol and chlorinated hydrocarbons. Immiscible with water, methanol and ethylene glycol.

Notes
Viscosity at 40cSt. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63148-62-9
Molecular Formula (C2H6OSi)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00132673
IUPAC Name Polydimethylsiloxane
SMILES C[Si](C)(-*)O-*

Specifications

Density 0.963
Flash Point >270°C (518°F)
Refractive Index 1.404
Quantity 100 g
Solubility Information Miscible with aliphatic,aromatic hydrocarbons,alcohol and chlorinated hydrocarbons. Immiscible with water,methanol and ethylene glycol.
Chemical Name or Material Silicone Oil
RTECSNumber JT6484500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.