Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silicone Oil Ap 100

Catalog No. 10838500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Click to view available options
Quantity:
100 mL
500 mL

Catalog No. 10-838-500ML

Supplier: Sigma Aldrich 10838500ML

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silicone oil AP 100 also known as polyphenyl-methylsiloxane (PPMS) is a high viscosity silicone compound that can be prepared by the anionic polymerization process. It has a polydispersity of 1.04, glass transition temperature (Tg) of -50C and surface tension of 27 nN/m.Silicone oil AP 100 can be potentially used as a carbon paste binder for the formation of tyrosinase electrode for biosensor applications. It forms a thin layer that facilitates the micro extraction of drugs from human breast milk.

Specifications

CAS 63148-58-3
Density 1.060 to 1.070 g/mL at 20°C
Refractive Index n20/D 1.502
Quantity 500 mL
MDL Number MFCD00132673
Synonym Polyphenyl-methylsiloxane
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.