Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Silicone Oil Ar 20
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
100 mL
500 mL
Description
Silicone oil AR 20 has been used as a segmenting fluid in polymerase chain reaction (PCR). It can be used in hollow fiber liquid-phase microextraction (LPME) procedure for extraction of hydrophobic drugs from human breast milk. It can also be used as a fluid to demonstrate pores in droplet interface bilayers (DIBs).
Specifications
Specifications
| CAS | 63148-58-3 |
| Density | 1.000 to 1.020 g/mL at 20°C |
| Refractive Index | n20/D 1.441-1.445 |
| MDL Number | MFCD00148386 |
| Quantity | 100 mL |
| Synonym | Polyphenyl-methylsiloxane |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction