Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific Silicone Sealing Mats for Deep Well Plates, 96 Square

Catalog No. 01183297 Shop All Simport Scientific Products
Change view
Click to view available options
Pre-slit:
No
Material:
Polyolefin, Rubber
Silicone
Pierceable:
No
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pre-slit Material Pierceable
01-183-297 No Polyolefin, Rubber No
01-183-298 No Silicone No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 01-183-297 Supplier Simport™ Scientific Supplier No. T11026
Add to Cart
Add to Cart
This item is not returnable. View return policy

Impermeable sealing mats minimize evaporation in a wide range of temperatures in PCR and storage applications. These silicone mats are chemical-resistant, reusable, and provide a high degree of confidence.

  • Effectively seals deep well plates
  • Easy to open and close
  • Prevents the volatilization of a variety of solvents
  • Chemical and heat resistant
  • All pre-slit models allow the pipette tip to enter the hole without damage to the silicone cover
  • Resistant to DMSO and biological buffers
  • Free of RNase, DNase, Pyrogen, and DNA

Specifications

Color Natural
Solvent Or Alcohol Resistant Alcohol Resistant, Solvent Resistant
Material Polyolefin, Rubber
Sterility Non-sterile
Autoclavable Autoclavable
No. of Wells 96
Pre-slit No
Certified RNase/DNase-free Yes
Pierceable No
Temperature Range (Metric) -80°C to +80°C
Well Shape Square
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.