Learn More
Bio Plas Siliconized Screw Cap Microcentrifuge Tubes
Catalog No. 0554161
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Capacity (Metric):
0.5 mL
1.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Capacity (Metric)
05-541-61 Sterile 0.5 mL
05-541-62 Non-sterile 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Siliconized products should NOT be autoclaved.
Catalog No. 05-541-61 Supplier Bio Plas Supplier No. 4200SLS
Siliconized products should NOT be autoclaved.
Ideal for packaging, transporting and storage of any diagnostic tests, especially in the area of biotechnology, biological products and reagents.
Extremely low surface adhesion, which is ideal for protein or nucleic acid studies to prevent any possible loss of sample due to adherence to the inner walls of the tube allowing complete sample recovery.
  • Positive screw cap seal with an O-ring designed to provide a safe, reliable secure fit even for repeated cap closures
  • Can withstand centrifugal forces up to 25,000xG
  • Conical, polypropylene, natural, available sterile or nonsterile
  • A tight seal closure eliminates accidental opening during incubation, spinning or transporting
Order Info
Caps are sold separately (Fisher Cat No. 05-541-70
Specifications
Closure Material Silicone
Autoclavable Not Autoclavable
Capacity (Metric) 0.5 mL
Color Natural
Material Polypropylene
Sterility Sterile
Graduated No
Closure Type Screw Cap
Bottom Shape Skirted
Max. RCF 25,000 x g