Learn More
Trajan Scientific and Medical™ SilTite™ Column Connector Kit
SDP
Catalog No. 073205 Shop All SGE Inc. Products
Change view
Click to view available options
Description:
0.32 mm I.D. column
Catalog No. Description
07-320-5 0.32 mm I.D. column
1 options
Catalog No. 07-320-5 Supplier Trajan Scientific and Medical™ Supplier No. 073205
Ensures easy, leak-free installation for capillary columns without the use of any tools.
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Type SilTite Kit 0.32 mm ID Column for Shimadzu
Description 0.32 mm I.D. column
Includes 10 ferrules, 2 nuts