Learn More
Trajan Scientific and Medical™ SilTite™ Column Connectors
SDP
Catalog No. 073273 Shop All SGE Inc. Products
Change view
Click to view available options
For Use With (Equipment):
1/32 in. Column Agilent
1/32 in. Column PerkinElmer
Shimadzu
Thermo Scientific
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
07-327-3 1/32 in. Column Agilent
07-323-3 Shimadzu
07-323-0 Thermo Scientific
07-321-9 1/32 in. Column PerkinElmer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07-327-3 Supplier Trajan Scientific and Medical™ Supplier No. 073273
Ensures easy, leak-free installation for capillary columns without the use of any tools.
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Type SilTite Kit 1/32 in. Column for Agilent
Description 1/32 in. column size
For Use With (Equipment) 1/32 in. Column Agilent