Learn More
Trajan Scientific and Medical™ SilTite™ Ferrules
SDP
Catalog No. 073229 Shop All SGE Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Shimadzu Detector/Injectors, Standard MS Interface 0.32 mm I.D. Columns
Shimadzu Detector/Injectors, Standard MS Interface 0.45 to 0.53 mm I.D. Columns
Thermo GC/MS Interface Connections 0.10 to 0.25 mm I.D. Columns
Thermo GC/MS Interface Connections 0.32 mm I.D. Columns
Thermo GC/MS Interface Connections 0.45 to 0.53 mm I.D. Columns
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
073229 Shimadzu Detector/Injectors, Standard MS Interface 0.45 to 0.53 mm I.D. Columns
073228 Shimadzu Detector/Injectors, Standard MS Interface 0.32 mm I.D. Columns
03-350-610 Thermo GC/MS Interface Connections 0.45 to 0.53 mm I.D. Columns
03-350-609 Thermo GC/MS Interface Connections 0.32 mm I.D. Columns
03-350-608 Thermo GC/MS Interface Connections 0.10 to 0.25 mm I.D. Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 073229 Supplier Trajan Scientific and Medical™ Supplier No. 073229
Ensures easy, leak-free installation for capillary columns without the use of any tools.
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
For Use With (Equipment) Shimadzu Detector/Injectors, Standard MS Interface 0.45 to 0.53 mm I.D. Columns
Type SilTite Ferrules for 0.53 mm ID Columns for Shimadzu