Learn More
Trajan Scientific and Medical™ SilTite™ Metal Nuts
SDP
Catalog No. 073232 Shop All SGE Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Shimadzu GC14 A, GC17 A, GC2010, GC2014 Detector/Injectors
Catalog No. For Use With (Equipment)
07-323-2 Shimadzu GC14 A, GC17 A, GC2010, GC2014 Detector/Injectors
1 options
Catalog No. 07-323-2 Supplier Trajan Scientific and Medical™ Supplier No. 073232
SilTite FingerTite delivers an easy, leak-free installation for capillary columns without the use of any tools
  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
For Use With (Equipment) Shimadzu GC14 A, GC17 A, GC2010, GC2014 Detector/Injectors
Type Nut
Material SilTite
Quantity 5/Pk.