Learn More
Eisco™ Siltstone Specimen
SCI_ED
Catalog No. S24745 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S24745 12/Pk.
S24744 1 Ea.
S26642 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S24745 Supplier Eisco™ Supplier No. ESNG0039PK12
Add to Cart
Add to Cart
Siltstone Specimen is a clastic rock made up of small, nearly microscopic grains of silt.
  • Clastic sedimentary rock made up of fine silt particles
  • For use in identifying depositional environments
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Clast size is indicative of low-energy environment
  • For clasts such as these to be deposited and lithified, they must be in environment with slowly moving or still water
Specifications
Product Type Sedimentary Rock
For Use With (Application) Identifying depositional environments
Packaging Clear labeled and barcoded bag
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Specimen Type Siltstone