Learn More
Silver 2,4-pentanedionate, 98%
Description
Silver 2,4-pentanedionate is used to make silver impregnated nanocrystalline metal oxides, with special emphasis on aluminum oxide. It is used as a cationization reagent in combination with polar matrices such as 2,5-dihydroxybenzoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Silver 2,4-pentanedionate is used to make silver impregnated nanocrystalline metal oxides, with special emphasis on aluminum oxide. It is used as a cationization reagent in combination with polar matrices such as 2,5-dihydroxybenzoic acid.
Solubility
Soluble in water.
Notes
Air, light and moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 100°C (decomposition) |
| Quantity | 5 g |
| Solubility Information | Soluble in water. |
| Formula Weight | 206.98 |
| Percent Purity | 98% |
| Odor | Characteristic |
| Sensitivity | Air, light, and moisture sensitive |
| Chemical Name or Material | Silver 2,4-pentanedionate |
Safety and Handling
| Hazard Category | H302+H312+H332-H315-H319-H335-H351 |
| Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 239-568-1 |
| TSCA | No |
| Recommended Storage | Keep cold; Store under Argon |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.