Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Silver
SCI_ED

Catalog No. S25522 Shop All Fisherbrand Products
Click to view available options
:
25g

Specifications

Color Silver
CAS 7440-22-4
Quantity 25 g
Assay Percent Range 1
Molecular Formula Ag
Linear Formula Ag
MDL Number MFCD00003397
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
SMILES [Ag]
IUPAC Name silver
Molecular Weight (g/mol) 107.87
PubChem CID 23954
ChEBI CHEBI:9141
Formula Weight 107.87
Packaging Foil
Density 10.5g/mL
Chemical Name or Material Silver
Show More Show Less
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.