Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver arsenate, 99.99% (metals basis), Thermo Scientific™

Catalog No. p-7059231
Click to view available options
Quantity:
5 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13510-44-6
Molecular Formula Ag3AsO4
Molecular Weight (g/mol) 462.522
MDL Number MFCD00046163
InChI Key IMGNYAPMSDUASV-UHFFFAOYSA-K
Synonym silver arsenate, unii-798q5px67i, trisilver arsenate, silberarsenat, trisilver arsorate, 3ag.aso4, arsenic acid h3aso4 , trisilver 1+ salt, arsenic acid h3aso4 , silver 1+ salt 1:3, the arsenic acid h3aso4 ,silver 1+ salt 1:3
PubChem CID 166835
IUPAC Name trisilver;trioxido(oxo)-$l^{5}-arsane
SMILES [O-][As](=O)([O-])[O-].[Ag+].[Ag+].[Ag+]

Specifications

Color Red
Physical Form Lump/Powder
Quantity 5 g
UN Number UN1557
Formula Weight 462.53
Percent Purity 99.99%
Sensitivity Light sensitive
Assay (metals basis)
Chemical Name or Material Silver arsenate
Hazard Category H301+H331-H350
Hazard Statement GHS H Statement
H301-H331-H350
Toxic if swallowed.
Toxic if inhaled.
May cause cancer.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
EINECSNumber 236-841-7
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.