Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Arsenite , in Sulfuric Acid, LabChem™

Catalog No. LC224222
Click to view available options
Quantity:
1 L
Packaging:
Amber Glass

Chemical Identifiers

CAS 7732-18-5, 7761-88-8, 7664-93-9, 1327-53-3
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide

Specifications

Color Colorless
CAS Max % 93.7, 3, 2, 1.3
Physical Form Liquid
Quantity 1 L
Linear Formula Ag3AsO3
UN Number UN2922
Solubility Information Soluble in water
Formula Weight 446.52
Identification Passes Test
Packaging Amber Glass
Decomposition Information Nitrogen oxides
Chemical Name or Material Silver Arsenite, in Sulfuric Acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Store locked up.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If in skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.