Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver bromide, 99.5%

Catalog No. p-4782119
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 7785-23-1 | AgBr | 187.772 g/mol

Silver bromide is used as the light-sensitive component on ordinary photographic film and plates and also used in I.R. applications and ionic semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7785-23-1
Molecular Formula AgBr
Molecular Weight (g/mol) 187.772
MDL Number MFCD00003398
InChI Key ADZWSOLPGZMUMY-UHFFFAOYSA-M
Synonym silver bromide, bromargyrite, bromyrite, nanoparticles, silver i bromide, silver bromide, premion
PubChem CID 66199
IUPAC Name bromosilver
SMILES Br[Ag]

Specifications

Melting Point 432°C
Color Yellow
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.9%
Solubility Information Insoluble in water,alcohol and acids. Partially soluble in ammonia.
Formula Weight 187.78
Odor Odorless
Sensitivity Light sensitive
Appearance Pale yellow powder/flake/crystalline/beads
Density 6.47 g/cm3 at 20°C
Chemical Name or Material Silver bromide
Show More Show Less
Hazard Category H400-H410: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
Hazard Statement Use appropriate containment to avoid environmental contamination. This material and/or its container must be disposed of as hazardous waste.
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 232-076-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.