Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver carbonate, 99.5% (metals basis)

Catalog No. AA1142022
Click to view available options
Quantity:
25 g
100 g

CAS: 534-16-7 | CH2Ag2O3 | 277.76 g/mol

Silver carbonate is used in the production of silver powder in microelectronics, conducting polymers, and in tooth surface treatment to prevent cavities. It is also used as an antimicrobial agent for infection resistant surgical fabric materials. It is used in organic synthesis, and in biological specimen staining.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-16-7
Molecular Formula CH2Ag2O3
Molecular Weight (g/mol) 277.76
MDL Number MFCD00003403
InChI Key WIKQEUJFZPCFNJ-UHFFFAOYSA-N
Synonym carbonic acid; silver, carbonic acid disilver
PubChem CID 25203940
IUPAC Name carbonic acid;silver
SMILES C(=O)(O)O.[Ag].[Ag]

Specifications

Melting Point 218°C (decomposition)
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Insoluble in water,acetone,acetate and alcohol. Freely soluble in dilute HNO3 and ammonia.
Formula Weight 275.75
Sensitivity Light sensitive
Density 6.08 g/mL
Chemical Name or Material Silver carbonate
Hazard Category H318
Precautionary Statement P280-P305+P351+P338-P310
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 208-590-3
RTECSNumber FG0700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.