Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver carbonate on Celite, ≈0.7 mmole Ag2CO3/g reagent

Catalog No. p-7049677
Click to view available options
Quantity:
15 mmol
75 mmol

Oxidizing agent for alcohols, diols/triols, hydroquinones, and amines | CAS: 534-16-7 | CH2Ag2O3 | 277.76 g/mol

Silver carbonate is used in production of silver powder. It is used in microelectronics. Silver carbonate is used as a reagent in organic synthesis. It is employed in Fetizon oxidation, silver carbonate on celite acts as an oxidizing agent to form lactones from diols. It is also used in used in Koenigs-Knorr glycosylation. It is also employed to convert alkyl bromides into alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-16-7
Molecular Formula CH2Ag2O3
Molecular Weight (g/mol) 277.76
MDL Number MFCD00003403
InChI Key WIKQEUJFZPCFNJ-UHFFFAOYSA-N
Synonym carbonic acid; silver, carbonic acid disilver
PubChem CID 25203940
IUPAC Name carbonic acid;silver
SMILES C(=O)(O)O.[Ag].[Ag]

Specifications

Physical Form Powder
Quantity 15 mmol
Solubility Information Soluble in water (0.03mg/ml at 20°C),dilute HNO3,ammonia and ammonium hydroxide. Insoluble in alcohol.
Odor Odorless
Sensitivity Light sensitive
Chemical Name or Material Silver carbonate on Celite, ∼0.7 mmole Ag2CO3/g reagent
Hazard Category H318-H335-H373
Hazard Statement GHS H Statement
H351-H373-H319
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
Precautionary Statement P260-P271-P280-P304+P340-P305+P351+P338-P310-P312-P314-P501c
DOTInformation Hazard Class: 9
EINECSNumber 208-590-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.