Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver casting grain, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4051130

Quantity 250g

 View more versions of this product

Catalog No. AA4051130


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7440-22-4
99.9% (Metals basis)
Silver
Casting Grain, 1.6-6.35mm (0.063-0.25 in.)
MFCD00003397

Specifications

250g
Silver-colored grain
Casting Grain, 1.6-6.35mm (0.063-0.25 in.)
MFCD00003397
Silver
7440-22-4
10.5g/cm3
99.9% (Metals basis)
Characteristic
Silver

Safety and Handling

SDS & Safety Documents

EINECSNumber : 231-131-3

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit