Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver chloride, 4.9% in H{2}O, nanoparticle dispersion, <200nm APS Number Weighted

Catalog No. p-7052366
Click to view available options
Quantity:
100 g
500 g

CAS: 7783-90-6 | AgCl | 143.32 g/mol

Used as pharmaceutical intermediate and in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-90-6
Molecular Formula AgCl
Molecular Weight (g/mol) 143.32
MDL Number MFCD00003399
InChI Key HKZLPVFGJNLROG-UHFFFAOYSA-M
Synonym Silver chloride dispersion in water
IUPAC Name silver(1+) chloride
SMILES [Cl-].[Ag+]

Specifications

Color White
Density 1.038 g/mL
Quantity 100 g
Odor Odorless
Particle Size <200nm APS Number Weighted
Sensitivity Light sensitive
Form Liquid
UN Number UN3082
Merck Index 14,8509
Solubility Information Dispersible in water. Very slightly soluble in cold water,soluble in solutions of alkali cyanides,thiosulfates,ammonium carbonates. Appreciably soluble in concentrated aqueous solutions of ammonium chloride,mercuric nitrate,and silver nitrate.
Formula Weight 143.32
Concentration or Composition (by Analyte or Components) 4.9% in H2O
Chemical Name or Material Silver chloride, Nanoparticle dispersion
Show More Show Less
Hazard Category H290
Precautionary Statement P234-P390
DOTInformation Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 232-033-3
RTECSNumber VW3563000
TSCA Yes
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.