Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver chloride, 99.9% (metals basis)

Catalog No. p-4407806
Click to view available options
Quantity:
25 g
100 g
500 g

Employed in Silver plating. Owing to its characteristic of reversible reduction to silver metal, it is used in photochromic lenses. Used as a cathode in sea water activated batteries. In electrochemistry, silver chloride electrode is used for potentiometric measurements. | CAS: 7783-90-6 | AgCl | 143.32 g/mol

Employed in Silver plating. Owing to its characteristic of reversible reduction to silver metal, it is used in photochromic lenses. Used as a cathode in sea water activated batteries. In electrochemistry, silver chloride electrode is used for potentiometric measurements. It serves as an antidote for mercury poisoning, and eliminates mercury from body. It is used in glass manufacturing industry. It is useful in the production of bandages, wound healing products and inglaze lustre, personal deodorant products, as well as for long term preservation of drinking water in water tanks; its pharmaceutical composition finds use as an antibacterial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-90-6
Molecular Formula AgCl
Molecular Weight (g/mol) 143.32
MDL Number MFCD00003399
InChI Key HKZLPVFGJNLROG-UHFFFAOYSA-M
IUPAC Name silver(1+) chloride
SMILES [Cl-].[Ag+]

Specifications

Boiling Point 1550°C
Melting Point 455°C
Physical Form Crystalline
Quantity 25 g
UN Number UN3077
Solubility Information Practically insoluble in water. Soluble in ammonia,solutions of alkali cyanides,thiosulfates and ammonium carbonates. Insoluble in alcohol and dilute acids.
Formula Weight 143.32
Percent Purity 99.9%
Packaging Glass bottle
Refractive Index 2.071
Sensitivity Light sensitive
Specific Gravity 5.56
Density 5.56
Chemical Name or Material Silver chloride
Show More Show Less
Hazard Category H290
Precautionary Statement P234-P390
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 232-033-3
RTECSNumber VW3563000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.