Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Silver chloride, 99+%, extra pure, ACROS Organics™
Click to view available options
Quantity:
10g
50g
250g
1kg
Packaging:
Glass bottle
Specifications
Assay Percent Range | 99+% |
Linear Formula | AgCl |
Fieser | 05,580; 10,347 |
Merck Index | 15,8648 |
Solubility Information | Solubility in water: 1.93mg/l (25°C). Other solubilities: insoluble in alcohol |
Formula Weight | 143.32 |
Physical Form | Powder |
Percent Purity | 99+% |
Grade | Extra Pure |
Copper (Cu) | 10ppm max. |
Show More |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Absorb spillage to prevent material damage.
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.
GHS Signal Word: Warning
EINECSNumber : 232-033-3