Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Chromate 99%, Thermo Scientific™

Catalog No. p-221897
Click to view available options
Quantity:
10 g
50 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-01-2
Molecular Formula Ag2CrO4
Molecular Weight (g/mol) 331.73
InChI Key OJKANDGLELGDHV-UHFFFAOYSA-N
Synonym silver chromate, unii-zj5092lwu9, silver chromate vi, silberchromat, silver i chromate, 2ag.cro4, chromic acid h2cro4 , disilver 1+ salt, chromic acid h2cro4 , silver 1+ salt 1:2, chromic acid, disilver i salt, disilver dioxido dioxo chromium
PubChem CID 62666

Specifications

Color Black to Gray
Physical Form Powder
Quantity 10 g
Linear Formula Ag2CrO4
Fieser 01,1005
Merck Index 15, 8649
SMILES [O-][Cr](=O)(=O)[O-].[Ag+].[Ag+]
IUPAC Name disilver;dioxido(dioxo)chromium
Formula Weight 331.72
Percent Purity ≥98.5% (AG2CRO4)
Density 5.62
Chemical Name or Material Silver chromate, 99%
Show More Show Less
Hazard Category HYGROSCOPIC
Hazard Statement LIGHT SENSITIVE
EINECSNumber 232-043-8
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.