Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver cyanate, 98+%

Catalog No. p-4510234
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle

CAS: 3315-16-0 | CAgNO | 149.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3315-16-0
Molecular Formula CAgNO
Molecular Weight (g/mol) 149.89
InChI Key VAQVDZZAAMAZAO-UHFFFAOYSA-N
Synonym silver,cyanate, cyanic acid; silver
PubChem CID 45051745
IUPAC Name cyanic acid;silver
SMILES C(#N)O.[Ag]

Specifications

Boiling Point 1085.0°C
Melting Point 652.0°C
Color Beige to Gray
Physical Form Powder
Quantity 10 g
Assay Percent Range 98% min
Linear Formula AgOCN
Fieser 02,224; 03,162
Solubility Information Solubility in water: soluble
Formula Weight 149.89
Percent Purity ≥98%
Packaging Glass bottle
Chemical Name or Material Silver cyanate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a
EINECSNumber 222-006-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.