Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silver hexafluoroantimonate(V)

Catalog No. 2277305G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-773-05G 5 g
22-773-025G 25 g
22-773-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-773-05G Supplier Sigma Aldrich Supplier No. 2277305G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Generates stable cation radical salts and is used as an acidic catalyst in epoxide opening reactions and sulfenylation reactions. Used to prepare silver 3,3G-dicyanodiphenylacetylene coordination networks for study of the effect of ligands on network structure toward the goal of engineering novel materials.

Specifications

CAS 26042-64-8
Molecular Formula AgF6Sb
Linear Formula AgSbF6
MDL Number MFCD00003401
Quantity 5 g
Molecular Weight (g/mol) 343.62
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.