Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Silver hexafluoroarsenate, 98+%

Click to view available options
Quantity:
10g
12005-82-2
AgAsF6
296.78
MFCD00014142
UNSKNPXMHZPWPB-UHFFFAOYSA-N
silver hexafluoroarsenate,silver hexafluoroarsenate v,silver hexafluoroarsoranuide,acmc-1c478,silver hexakis fluoranyl arsenic 1-,silver 1+ hexafluoro-??-arsanuide,silver 1+ ion hexafluoro-??-arsanuide
11077325
silver;hexafluoroarsenic(1-)
F[As-](F)(F)(F)(F)F.[Ag+]
12005-82-2
AgAsF6
296.78
MFCD00014142
UNSKNPXMHZPWPB-UHFFFAOYSA-N
silver hexafluoroarsenate,silver hexafluoroarsenate v,silver hexafluoroarsoranuide,acmc-1c478,silver hexakis fluoranyl arsenic 1-,silver 1+ hexafluoro-??-arsanuide,silver 1+ ion hexafluoro-??-arsanuide
11077325
silver;hexafluoroarsenic(1-)
F[As-](F)(F)(F)(F)F.[Ag+]
UN Number UN1557
Formula Weight 296.78
Physical Form Powder
Percent Purity ≥98%
Odor Odorless
Sensitivity Moisture sensitive
Color White
Quantity 10g
Chemical Name or Material Silver hexafluoroarsenate

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures