Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver(I) fluoride, 99+%

Catalog No. p-221925
Click to view available options
Quantity:
5 g
25 g
Packaging:
Plastic bottle

CAS: 7775-41-9 | AgF | 126.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7775-41-9
Molecular Formula AgF
Molecular Weight (g/mol) 126.87
MDL Number MFCD00003410
InChI Key REYHXKZHIMGNSE-UHFFFAOYSA-M
Synonym silver fluoride, silver 1+ fluoride, silver 1+ ion fluoride, argentous fluoride, ag-f, silverfluoride, silver salt, fluoride, silver fluoride,, silver i fluoride 5g
PubChem CID 165912
ChEBI CHEBI:30340
IUPAC Name silver;fluoride
SMILES [F-].[Ag+]

Specifications

Boiling Point 1150.0°C
Melting Point 435.0°C
Color Brown-Rust to Brown-Yellow
Physical Form Powder
Quantity 5 g
Assay Percent Range 99% min
Linear Formula AgF
Fieser 02,364; 05,581; 06,514; 15,287
Merck Index 15, 8653
Solubility Information Solubility in water: in water: 182 g/100mL (15.5°C).
Formula Weight 126.87
Percent Purity ≥99%
Packaging Plastic bottle
Specific Gravity 5.85
Density 5.8500g/mL
Chemical Name or Material Silver(I) fluoride, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 231-895-8
RTECSNumber VW4250000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.