Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver(I) oxide, 99.99% (metals basis)

Catalog No. aa4257703
Click to view available options
Quantity:
1 g
5 g
25 g
4 x 25 g

CAS: 20667-12-3 | Ag2O | 231.74 g/mol

Polishing glass, coloring glass yellow, catalyst, purifying drinking water, lab reagent, carbon dioxide scrubber, and chemical sensors. It is used in the preparation of other silver compounds, and silver-oxide batteries. In organic chemistry, silver oxide finds use as an oxidizing agent for aldehyes to produce carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20667-12-3
Molecular Formula Ag2O
Molecular Weight (g/mol) 231.74
MDL Number MFCD00003404
InChI Key KHJDQHIZCZTCAE-UHFFFAOYSA-N
IUPAC Name (argentiooxy)silver
SMILES [Ag]O[Ag]

Specifications

Melting Point 230°C (decomposition)
Color Brown
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in dilute nitric acid and ammonia. Practically insoluble in water and alcohol
Formula Weight 231.74
Odor Odorless
Sensitivity Light sensitive
Appearance Dark brown powder
Density 7.2
Chemical Name or Material Silver(I) oxide
Show More Show Less
missing translation for 'healthHazard1' H271-H318
missing translation for 'healthHazard3' P210-P220-P221-P280-P283-P305+P351+P338-P306+P360-P310-P370+P378q-P371+P380+P375-P501c
DOTInformation DOT Class: 5.1; Packaging Group: I
EINECSNumber 243-957-1
RTECSNumber VW4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.