Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver(I) oxide, Electrical Grade

Catalog No. p-7058793
Click to view available options
Quantity:
250 g
1 kg

For battery plates, pollution control filters | CAS: 20667-12-3 | Ag2O | 231.74 g/mol

For battery plates, and pollution control filters. It is used in the preparation of other silver compounds, and silver-oxide batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20667-12-3
Molecular Formula Ag2O
Molecular Weight (g/mol) 231.74
MDL Number MFCD00003404
InChI Key KHJDQHIZCZTCAE-UHFFFAOYSA-N
IUPAC Name (argentiooxy)silver
SMILES [Ag]O[Ag]

Specifications

Melting Point 230°C (decomposition)
Color Brown
Physical Form Powder
Quantity 250 g
UN Number UN1479
Merck Index 14,8521
Solubility Information Soluble in dilute HNO3 and ammonia; Practically insoluble in water; Insoluble in alcohol
Formula Weight 231.74
Grade Electronic
Odor Odorless
Sensitivity Light sensitive
Density 7.2 g/mL
Chemical Name or Material Silver (I) oxide
Show More Show Less
Hazard Category H271-H318
Hazard Statement GHS H Statement
H271-H318
May cause fire or explosion
strong oxidizer.
Causes serious eye damage.
Precautionary Statement P210-P220-P221-P280-P283-P305+P351+P338-P306+P360-P310-P370+P378q-P371+P380+P375-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: I
EINECSNumber 243-957-1
RTECSNumber VW4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.