Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver(II) fluoride, 98+%

Catalog No. aa1161004
Click to view available options
Quantity:
2 g
10 g
50 g

Moisture and Light Sensitive | CAS: 7783-95-1 | AgF2

Silver(II) fluoride is used as a fluorinating and oxidizing agent. It is also used in the preparation of organic perfluorocompounds and in the fluorination of aromatic compounds. In addition, it is also used in pharmacy and electroplating industry. It serves as an intermediate in the catalysis of gaseous reactions with fluorine by silver. It is utilized in the oxidation of xenon to xenon difluoride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-95-1
Molecular Formula AgF2
MDL Number MFCD00003411
Synonym Silver difluoride

Specifications

Melting Point 690°C
Color Brown
Physical Form Crystalline
Quantity 2 g
Assay Percent Range 98+%
Solubility Information Soluble in hexane,chloroform,chlorofluorocarbon and hydrogen fluoride.
Formula Weight 145.87
Sensitivity Light and moisture sensitive
Appearance Brown crystals
Density 4.6
Chemical Name or Material Silver(II) fluoride
Show More Show Less
Hazard Category H272-H302+H312+H332-H314-H335
Precautionary Statement P210-P220-P221-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 232-037-5
RTECSNumber VW4200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.