Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver(II) oxide, 85.5-88.0% Ag, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
1301-96-8
AgO
123.87
MFCD00044285
OTCVAHKKMMUFAY-UHFFFAOYSA-N
silver ii oxide, silver oxide, silberperoxyd, silver oxide,ago, acmc-1aiyc, silver i,iii oxide, ksc203c6r, silver ii oxide 5g, silver ii oxide-ag
92152
oxosilver
O=[Ag]
1301-96-8
AgO
123.87
MFCD00044285
OTCVAHKKMMUFAY-UHFFFAOYSA-N
silver ii oxide, silver oxide, silberperoxyd, silver oxide,ago, acmc-1aiyc, silver i,iii oxide, ksc203c6r, silver ii oxide 5g, silver ii oxide-ag
92152
oxosilver
O=[Ag]
Linear Formula AgO
Fieser 02,369; 04,431; 06,518; 07,322; 09,412; 10,352; 11,469
Merck Index 15, 8661
Solubility Information Solubility in water: insoluble in water
Formula Weight 123.87
Physical Form Powder
Percent Purity 85.5-88% Ag
Packaging Glass bottle
Color Black to Gray
Melting Point >100°C
Quantity 10g
Chemical Name or Material Silver(II) oxide, 85.5-88.0% Ag
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 215-098-2

TSCA : TSCA