Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Silver(II) oxide, 98%

Manufacturing of silver oxide-zinc alkali batteries

Manufacturer:  Alfa Aesar™ 2269506

Quantity 5g

 View more versions of this product

Catalog No. AA2269506


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1301-96-8
MFCD00044285
14,8522
Silver(II) oxide
123.87
UN3085
Silver peroxide; Silver suboxide

Specifications

1301-96-8
123.87
UN3085
Silver peroxide; Silver suboxide
7.44g/mL
Moisture & Light Sensitive
>100°C (dec.)
Silver(II) oxide
Powder
MFCD00044285
14,8522
Insoluble in water; Soluble in alkalies, ammonium hydroxide with decomposition and N2 formation
Odorless
Gray to black
5g
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H272-H314-H318
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit