Learn More
Silver(II) oxide, 99.9% (metals basis), Ag 86.6% min
Description
Silver(II) oxide is used to make silver oxide-zinc alkali batteries. It is also used as a reagent in laboratory reactions and forms transition metal-carbene complexes. It is also used as a catalyst to prepare silver metal. It acts as an effective carbon dioxide scrubber and used in nuclear submarines, the international space station and space shuttles. Further, it is used in the manufacture of filters for gas sensors and in some infection-resistant surgical fabric materials and fibrous textile articles. In addition to this, it is used in concrete and in some swimming pools to protect the water from undesired microbes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | >100°C (decomposition) |
Color | Black to Gray |
Physical Form | Powder |
Quantity | 5 g |
UN Number | UN3085 |
Merck Index | 14,8522 |
Solubility Information | Soluble in alkalis. Insoluble in water. |
Formula Weight | 123.87 |
Percent Purity | 99.9% |
Concentration or Composition (by Analyte or Components) | Ag 86.6% min. |
Show More |
Safety and Handling
Hazard Category | H272-H314-H318-H335 |
Hazard Statement | GHS H Statement H272-H314-H318 May intensify fire oxidizer. Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P370+P380+P375-P372-P380-P501c |
DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S. |
EINECSNumber | 215-098-2 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.