Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver iodide, 99%, pure, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
7783-96-2
AgI
234.77
MFCD00003412
MSFPLIAKTHOCQP-UHFFFAOYSA-M
silver iodide,neosiluol,neosilvol,silver monoiodide,silver 1+ iodide,colloidal silver iodide,iodanylsilver,silver iodide metals basis,ccris 8490
24563
iodosilver
[Ag]I
7783-96-2
AgI
234.77
MFCD00003412
MSFPLIAKTHOCQP-UHFFFAOYSA-M
silver iodide,neosiluol,neosilvol,silver monoiodide,silver 1+ iodide,colloidal silver iodide,iodanylsilver,silver iodide metals basis,ccris 8490
24563
iodosilver
[Ag]I
Assay Percent Range 98.5% min. (ex Ag)
Linear Formula AgI
Merck Index 15, 8655
Solubility Information Solubility in water: 0.03mg/l
Formula Weight 234.77
Physical Form Granular Powder
Percent Purity 99%
Grade Pure
Density 5.6800g/mL
Packaging Glass bottle
Specific Gravity 5.68
Boiling Point 1506°C
Color Yellow
Melting Point 557°C
Quantity 5g
Chemical Name or Material Silver iodide, Pure
Show More Show Less

GHS H Statement
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.

GHS Signal Word: Warning

EINECSNumber : 232-038-