Learn More
Silver lactate
Description
Silver lactate is used as a surgical antiseptic. It is also used as an antimicrobial agent in the external applications. Further, it is used in the treatment of full thickness thermal wounds. In addition to this, it is employed as a hypodermic injection in erysipelas
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 120°C to 122°C |
| Color | White to Gray |
| Physical Form | Powder |
| Quantity | 10 g |
| Linear Formula | CH3CH(OH)CO2Ag |
| UN Number | UN3077 |
| Merck Index | 14,8517 |
| Solubility Information | Soluble in water. Slightly soluble in alcohol. |
| Formula Weight | 196.94 |
| Odor | Odorless |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H315 Harmful if swallowed. Causes skin irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| RTECSNumber | UA2465550 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.