Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Silver lactate, 96%
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 120.0°C to 122.0°C |
Color | Brown-Gray to Gray |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 95% min. (Argentometry) |
Linear Formula | CH3CH(OH)CO2Ag |
Merck Index | 14,8517 |
Solubility Information | Solubility in water: 77g/L (20°C). Other solubilities: slightly soluble in alcohol |
Formula Weight | 196.94 |
Percent Purity | 96% |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. If eye irritation persists: Get medical advice/attention. IF INHALED. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protectiv |
EINECSNumber | 239-859-3 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction