Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Silver nanoparticles, 80nm, 0.02mg/mL, supplied in 2mM sodium citrate, 457nm absorption

Click to view available options
Quantity:
25mL
100mL
7440-22-4
Ag
107.868
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber
23954
CHEBI:9141
silver
[Ag]
7440-22-4
Ag
107.868
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber
23954
CHEBI:9141
silver
[Ag]
Applications
Silver nanoparticles applications include molecular diagnostics and photonic devices, which take advantage of the novel optical properties of these nanomaterials. An increasingly common application is the use of silver nanoparticles for antimicrobial coatings, and many textiles, keyboards, wound dressings, and biomedical devices now contain silver nanoparticles that continuously release a low level of silver ions to provide protection against bacteria.

Solubility
Fully miscible in water.

Notes
Light and air sensitive. Store away from heat, light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Quantity 25mL
Odor Odorless
Physical Form Liquid
Particle Size 80nm
Sensitivity Light Sensitive
Color Yellow
Chemical Name or Material Silver nanoparticles, 0.02mg/mL, supplied in 2mM sodium citrate, 457nm absorption

TSCA : TSCA

storageNote1 : 2° to 8°C