Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silver nanowires

Catalog No. 77809525ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
25 mL

Catalog No. 77-809-525ML

Supplier: Sigma Aldrich 77809525ML

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silver nanowires have been studied for use in electronic and biological applications including transparent conductors; and water sterilization . Silver nanowires have also been studied as plasmonic and as components in amorphous semiconductors.

Specifications

CAS 7440-22-4
Density 0.779 g/mL (at 25°C)
Molecular Formula Ag
Linear Formula Ag
Quantity 25 mL
MDL Number MFCD00003397
Synonym Silver nanofibers; Silver nanowhiskers; Silver nanowire
Molecular Weight (g/mol) 107.87
Biological Activity Central nervous system
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.