Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
For Chloride Titrations
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
20 L
Packaging:
Amber Poly Bottle
Cubitainer
Specifications
Specifications
Color | Colorless |
CAS Min % | 98.0, 0.47 |
CAS Max % | 100.0, 0.49 |
Grade | Laboratory |
Packaging | Amber Poly Bottle |
Solvent or Matrix | 1 mL = 1 mg Cl |
pH | ∼6 |
Quantity | 500 mL |
Physical Form | Liquid |
Concentration or Composition Notes | 0.0282Normal |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction